missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human LIMK1, His Tag Recombinant Protein
Brand: Invitrogen™ PV4348
Additional Details : Weight : 0.00945kg
Description
- LIMK1 is a serine/threonine kinase
- It is involved in regulation of actin cytoskeletal changes via phosphorylation of cofilin
- LIMK1 has also been implicated in brain development
Specifications
NP_002305 | |
3984 | |
1 mg | |
LISK Kinase Family | |
Recombinant | |
Storage Buffer: 50 mM Tris pH 7.5, 150 mM NaCl, 0.5 mM EDTA, 0.02% Triton X-100, 2 mM DTT, 50% glycerol Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
Dry Ice | |
Kinases & Inhibitors | |
Frozen |
Kinase Assay | |
44.7 kDa | |
LIMK1, LIMK | |
Human | |
His Tag | |
Ser/Thr Kinases: TKL Group | |
Baculovirus | |
Catalytic | |
Drug Development |