Få mere at vide
Novus Biologicals™ Lightning-Link (R) Rapid Atto633 Antibody Labeling Kit
Artikelnummer. 18030514 Shop alle Bio Techne produkter
Mængde:
100 μg
Artikelnummer. 18030514
Brand: Novus Biologicals™ 3530005
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Antibody labeling kit
Lightning-Link™ Rapid Antibody Labeling kits enable direct labeling of antibodies, proteins, peptides or other biomolecules and allow conjugations to be set up in seconds, and used within minutes (less than 20 minutes).
Kit Contents:
  • Lightning-Link™ Rapid mix
  • 1 vial of LL Rapid Quencher reagent
  • 1 vial of LL Rapid Modifier reagent
TRUSTED_SUSTAINABILITY
Tekniske data
Mængde 100 μg
Produkttype Antibody Labeling Kit
Indhold og opbevaring -20°C