Få mere at vide
Novus Biologicals™ Lightning-Link (R) Rapid Alexa Fluor 568 Antibody Labeling Kit
Artikelnummer. 18067343 Shop alle Bio Techne produkter
Mængde:
1 mg
100 μg
30 μg
300 μg
Artikelnummer. 18067343
Brand: Novus Biologicals™ 3340015
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Antibody labeling kit
Lightning-Link™ Rapid Antibody Labeling kits enable direct labeling of antibodies, proteins, peptides or other biomolecules and allow conjugations to be set up in seconds, and used within minutes (less than 20 minutes).
Kit Contents:
  • Lightning-Link™ Rapid mix
  • 1 vial of LL Rapid Quencher reagent
  • 1 vial of LL Rapid Modifier reagent
TRUSTED_SUSTAINABILITY
Tekniske data
Mængde 1 mg
Produkttype Antibody Labeling Kit
Indhold og opbevaring -20°C