Få mere at vide
Novus Biologicals™ Lightning-Link (R) Rapid Alexa Fluor 555 Antibody Labeling Kit
Shop alle Bio Techne produkter
Mængde:
1 mg
100 μg
30 μg
300 μg
Beskrivelse
Lightning-Link™ Rapid Antibody Labeling kits enable direct labeling of antibodies, proteins, peptides or other biomolecules and allow conjugations to be set up in seconds, and used within minutes (less than 20 minutes).
Kit Contents:
- 1 vial of LL Rapid Modifier reagent
- 1 vial of LL Rapid Quencher reagent
- Lightning-Link™ Rapid mix
Tekniske data
Tekniske data
| Mængde | 100 μg |
| Produkttype | Antibody Labeling Kit |
| Indhold og opbevaring | -20°C |