Få mere at vide
Novus Biologicals™ Lightning-Link (R) Fluorescein Antibody Labeling Kit
Shop alle Bio Techne produkter
Mængde:
100 μg
Beskrivelse
Lightning-Link™ antibody labeling kits enable the direct labeling of antibodies, proteins, peptides or other biomolecules for use in R&D applications, drug discovery and the development of diagnostic kits.
Kit Contents:
- 1 vial of LL-modifier reagent
- 1 vial of LL-quencher FD reagent
- 1 glass vial of Lightning-Link™ mix
Tekniske data
Tekniske data
| Mængde | 100 μg |
| Produkttype | Antibody Labeling Kit |
| Indhold og opbevaring | -20°C |