Learn More
Novus Biologicals™ Lightning-Link (R) APC-Cy7 Antibody Labeling Kit
Applications: Flow Cytometry
Brand:  Novus Biologicals™ 765-0030
 View more versions of this product
Product Code. 18759733
  • 4010.00 DKK / 30µg
Description
APC/Cy7 is a tandem conjugate. In the Lightning-Link APC/Cy7 the tandem is excited at 633nm, wavelength at which the APC has its maximum absorption. The excited APC functions then as an energy donor, where the energy is transferred via FRET (Fluorescence Resonance Energy Transfer) to the Cy7. The Cy7 emits then the energy received from the APC in the form of long wavelength light at 776nm.
Order Info
Quantity
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Store at -20°C. Avoid freeze-thaw cycles.
APC/Cy7
APC-Cy7 Antibody Labeling Kit
3 x 0.01 mg Reaction
Glass vial(s) of Lightning-Link mix (1 or 3 vials, depending on pack size), 1 vial of LL Modifier reagent, 1 vial of LL Quencher FD reagent
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Novus Biologicals™ Lightning-Link (R) APC-Cy7 Antibody Labeling Kit > 3 x 0.01mg Reactions