Få mere at vide
Invitrogen™ EVOS™ Light Cube, TagBFP
Shop alle Thermo Scientific produkter
Mængde:
1 Ea.
Pakningsstørrelse:
Hver
Beskrivelse
- Contains unique, proprietary LED light cubes
- Image fluorescent samples for specific wavelengthLight engine outputs high intensity over a short light path that delivers superior fluorophore excitation
- Each cube contains an LED, condensing optics, and hard-coated filters
- User-changeable and automatically recognized by system
- Shorter light path that affords superior detection of fluorescent signals
- Continuous illumination that enables consistent results
- 50,000+ hour bulb lifetime that affords lower operating costs
- Adjustable light intensity that helps reduce photobleaching
Tekniske data
Tekniske data
| Type | Light Cube |
| Produktlinje | EVOS |
| Til brug med (udstyr) | Countess™ II FL Automated Cell Counter, EVOS™ M5000 Imaging System, EVOS™ M7000 Imaging System |
| Inkluderer | Condensing Optics, Hard-Coated Filters, LED |
| Bølgelængde | Ex: 390/18 Em: 447/60 |
| Lyskilde | Metal Halide (Bulb) |
| Mængde | 1 Ea. |