Learn More
LIFR alpha Antibody (12D3), Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Bio-Techne NBP3-18054
Product Code. 18310474
  • 2320.00 DKK / 0.10mg
Description
LIFR alpha Monoclonal antibody specifically detects LIFR alpha in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
LIFR alpha
Monoclonal
1 mg/ml
Western Blot, Flow Cytometry 2-5 ug/ml, ELISA
CD118, CD118 antigen, FLJ98106, FLJ99923, leukemia inhibitory factor receptor, leukemia inhibitory factor receptor alpha, LIF receptor, LIF-R, SJS2, STWS, SWS
Human recombinant soluble LIFR alpha-Fc
0.1 mg
Immunology
3977
Store at 4C. Do not freeze.
IgG1 κ
Western Blot, Flow Cytometry, ELISA
12D3
Unconjugated
PBS, pH 7.4
Mouse
Protein A purified
RUO
Primary
Human
Purified