Få mere at vide
BOLA Liebig Kondensatorer Gennemsigtige
Artikelnummer. 10247351
Skift visning
Længde (metrisk) jakke:
450 mm
600 mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Længde (metrisk) jakke
10247351 450 mm
10247401 600 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 10247351 Leverandør BOLA Leverandørnr. B29104
Læg i indkøbskurv
Læg i indkøbskurv
Liebig-kondensatorer Transparent PTFE, med SAFE LAB-system
  • Thin-walled cooling tube made of translucent and gastight PFA
  • cooling jacket made of borosilicate glass with hose connectors
  • made of PP and nuts for connection of cooling water
  • ground joint cone and socket size 29 made of PTFE.
  • The distillate is only exposed to PFA/PTFE. Integrated special nut (Safe-Lab) for easy locking and unlocking of the ground joint.
Tekniske data
Certificeringer/compliance FDA
Længde (metrisk) jakke 450 mm