Få mere at vide
Lidoflazine, LoGiCal
Artikelnummer. 30457778
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30457778 10 mg
1 options
Artikelnummer. 30457778 Leverandør LoGiCal Leverandørnr. LGCFOR3120.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Lidoflazine
CAS 3416-26-0
Analyt- eller komponentnavne Lidoflazine
Forsendelsestilstand Room Temperature
Molekylær formel C30 H35 F2 N3 O
Certificeringer/compliance ISO 9001
Synonym 1-Piperazineacetamide, 4-[4,4-bis(4-fluorophenyl)butyl]-N-(2,6-dimethylphenyl)-, 1-Piperazineaceto-2',6'-xylidide, 4-[4,4-bis(p-fluorophenyl)butyl]- (7CI,8CI), 1-[4,4-Bis(4-fluorophenyl)butyl]-4-(2,6-dimethylphenylcarbamoylmethyl)piperazine, 1-[4,4-Bis(fluorophenyl)butyl]-4-(2,6-dimethylanilinocarbonylmethyl)piperazine, 1-[4,4-Di(4-fluorophenyl)butyl]-4-[(2,6-dimethylanilinocarbonyl)methyl]piperazine, 4-[4,4-Bis(p-fluorophenyl)butyl]-1-piperazineaceto-2',6'-xylidide, Angex, Clinium, Corflazine, Klinium, Lidoflazin, Lidoflazine, Lidoflazinum, McN-JR 7094, McN-JR 7904, Ordiflazine, R 7904
InChI formel InChI=1S/C30H35F2N3O/c1-22-5-3-6-23(2)30(22)33-29(36)21-35-19-17-34(18-20-35)16-4-7-28(24-8-12-26(31)13-9-24)25-10-14-27(32)15-11-25/h3,5-6,8-15,28H,4,7,16-21H2,1-2H3,(H,33,36)
Løsningstype Neat
SMIL Cc1cccc(C)c1NC(=O)CN2CCN(CCCC(c3ccc(F)cc3)c4ccc(F)cc4)CC2
IUPAC navn 2-[4-[4,4-bis(4-fluorophenyl)butyl]piperazin-1-yl]-N-(2,6-dimethylphenyl)acetamide
Industry Type Clinical Testing
Molekylvægt (g/mol) 491.62
Mængde 10 mg
Formel vægt 491.2748 g/mol