Få mere at vide
Lidocaine hydrochloride, MedChemExpress
Artikelnummer. 16468388
Encompass_Preferred
Skift visning
Mængde:
5 g
10 g
Pakningsstørrelse:
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
16468388 10 g -
16430808 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 16468388 Leverandør Medchem Express Leverandørnr. HYB0185A10g
MedChemExpress Lidocaine hydrochloride (Lignocaine hydrochloride) inhibits sodium channels involving complex voltage and using dependence.
Lidocaine hydrochloride decreases growth, migration and invasion of gastric carcinoma cells via up-regulating miR-145 expression and further inactivation of MEK/ERK and NF-κB signaling pathways. Lidocaine hydrochloride is an amide derivative and a drug to treat ventricular arrhythmia and an effective tumor-inhibitor.
Tekniske data
Kemisk navn eller materiale Lidocaine hydrochloride
Fysisk form Solid
CAS 73-78-9
Farve White
Mængde 10 g
Renhedsgrad noter Research
Molekylær formel C14H23ClN2O
Til brug med (applikation) Cancer-Kinase/protease
Synonym Lignocaine hydrochloride
Opløselighedsinformation H2O : ≥ 100 mg/mL (369.28 mM) ∣DMSO : ≥ 100 mg/mL (369.28 mM)
SMIL O=C(NC1=C(C)C=CC=C1C)CN(CC)CC.[H]Cl
Molekylvægt (g/mol) 270.8
Formel vægt 270.8
Procent renhed 99.46%