Få mere at vide
Lidocaine EP Impurity K (as Hydrochloride), Mikromol™
Beskrivelse
Lidocaine EP Impurity K (as Hydrochloride)
Tekniske data
Tekniske data
| Kemisk navn eller materiale | N-(2,6-Dimethylphenyl)-2-(ethylmethylamino)acetamide Hydrochloride |
| CAS | 50295-20-0 |
| Analyt- eller komponentnavne | N-(2,6-Dimethylphenyl)-2-(ethylmethylamino)acetamide Hydrochloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C13 H20 N2 O . Cl H |
| Certificeringer/compliance | ISO 17025 |
| Synonym | N-(2,6-Dimethylphenyl)-2-(ethylmethylamino)acetamide Hydrochloride, Lidocaine Hydrochloride Imp. K (EP) as Hydrochloride, Lidocaine Hydrochloride Imp. K (EP), Lidocaine Imp. K (EP), Lidocaine Hydrochloride Impurity K as Hydrochloride, Lidocaine Impurity K as Hydrochloride |
| InChI formel | InChI=1S/C13H20N2O.ClH/c1-5-15(4)9-12(16)14-13-10(2)7-6-8-11(13)3;/h6-8H,5,9H2,1-4H3,(H,14,16);1H |
| SMIL | Cl.CCN(C)CC(=O)Nc1c(C)cccc1C |
| IUPAC navn | N-(2,6-dimethylphenyl)-2-[ethyl(methyl)amino]acetamide;hydrochloride |
| Vis mere |