Få mere at vide
Greiner Bio-One Lid for 96-well Plate, Polystyrene, High Profile (9 mm), Sterile
Beskrivelse
- Clear, polystyrene construction
- Free of detectable DNase, RNase, human DNA
- Non-pyrogenic, non-cytotoxic
- Offer protection against contamination and evaporation during sample storage and cell cultivation
- Suitable for non-TC-treated 96-well microplates
- High lid profile; height: 9 mm
- Sterile
Tekniske data
Tekniske data
| Materiale | Polystyrene |
| Sterilitet | Sterile |
| nr. pr. sag | 100/Cs. |
| nr. pr. pakke | 1/Pk |
| Pladekompatibilitet | 96-brønds plade |
| Afskårne hjørner | Yes |