Få mere at vide
Grant Instruments™ Plastic Lid for Grant Stirred Water Baths
Til brug med (udstyr):
P12 Plastiktank
P18 Plastiktank
P5 Plastiktank
Pakningsstørrelse:
Hver
Beskrivelse
Helps to reduce evaporation/heat loss and avoid sample contamination. Grant Instruments™ Plastic Lid for Grant Stirred Water Baths is constructed of curved or flat plastic with an integral handle
- Flat or curved moulded plastic
- Integral handle
- For use with Grant Stirred Water Baths and heated Circulators
- Prevents heat from escaping
- Reduces sample contamination
Tekniske data
Tekniske data
| Oversigt | Lid for 12L tank |
| Til brug med (udstyr) | P12 Plastiktank |
| Materiale | Plastic |