missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LGALS3BP Monoclonal Antibody (D4), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA543994
This item is not returnable.
View return policy
Description
The galectins are a family of beta-galactoside-binding proteins implicated in modulating cell-cell and cell-matrix interactions. LGALS3BP has been found elevated in the serum of patients with cancer and in those infected by the human immunodeficiency virus (HIV). It appears to be implicated in immune response associated with natural killer (NK) and lymphokine-activated killer (LAK) cell cytotoxicity. Using fluorescence in situ hybridization the full length 90K cDNA has been localized to chromosome 17q25. The native protein binds specifically to a human macrophage-associated lectin known as Mac-2 and also binds galectin 1.
Specifications
LGALS3BP | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
Q08380 | |
LGALS3BP | |
Recombinant protein Lectin Galactoside Binding, Soluble 3 Binding Protein. The antigen corresponds to amino acid range 24-221 of the target protein. | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG2b κ |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
D4 | |
Unconjugated | |
LGALS3BP | |
90K; basement membrane autoantigen p105; BTBD17B; CyCAP; Cyp-C-associated protein; galectin 3 binding protein; galectin-3-binding protein; gp90; L3 antigen; Lectin; lectin galactoside-binding soluble 3 binding protein; lectin galactoside-binding soluble 3-binding protein; lectin, galactoside binding soluble 3 binding protein; lectin, galactoside-binding, soluble, 3 binding protein; Lgals3bp; M2BP; mac-2 BP; mac-2-binding protein; MAC2BP; MAC-2BP; MAC-2-BP; mama; peptidylprolyl isomerase C-associated protein; Ppicap; Protein 90K; Protein MAMA; serum protein 90K; similar to Galectin-3 binding protein precursor (Lectin galactoside-binding soluble 3 binding protein) (Mac-2 binding protein) (Mac-2 BP) (MAC2BP) (Tumor-associated antigen 90K); TANGO10B; transport and golgi organization 10 homolog B; tumor-associated antigen 90K | |
Mouse | |
Protein A/G | |
RUO | |
3959 | |
-20°C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
LGALS3BP Monoclonal Antibody (D4), Invitrogen™ > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction