Få mere at vide
LGALS3 (Human) Matched Antibody Pair, Polyclonal , Abnova™
Beskrivelse
Indeholder
Capture antibody: mouse monoclonal anti-LGALS3, IgG2a Kappa (100μg)
Detection antibody: rabbit purified polyclonal anti-LGALS3 (50μg) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
lectin, galactoside-binding, soluble, 3
Tekniske data
Tekniske data
| Antigen | LGALS3 |
| Applikationer | ELISA |
| Klassifikation | Monoclonal/Polyclonal |
| Konjugeret | Unconjugated |
| Oversigt | This antibody pair set comes with matched antibody pair to detect and quantify protein level of human LGALS3. |
| Sygdomstilstand | Carcinoma, Papillary Cardiovascular Diseases, Diabetes Mellitus Type 1, Diabetes Mellitus Type 2, Diabetic Nephropathies, Edema, Genetic Predisposition to Disease, Thyroid Neoplasms |
| Gene | LGALS3 |
| Gene Alias | CBP35/GAL3/GALBP/GALIG/LGALS2/MAC2 |
| Gensymboler | LGALS3 |
| Mængde | 1 Set |
| Vis mere |