Få mere at vide
Levulinic-d5 Acid, TRC
Artikelnummer. 30469960
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30469960 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30469960 Leverandør Toronto Research Chemicals Leverandørnr. TRCL37950225MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Levulinic-d5 Acid
CAS 1206185-52-5
Molekylær formel C5 D5 H3 O3
Mængde 25 mg
Synonym 3-Acetylpropionic-d5 Acid, 3-Oxobutanecarboxylic-d5 Acid, 4-Ketovaleric-d5 Acid, 4-Oxopentanoic-d5 Acid, 4-Oxovaleric-d5 Acid, Laevulinic-d5 Acid, Levulic-d5 Acid, NSC 3716, β-Acetylpropionic-d5 Acid, γ-Ketovaleric-d5 Acid, Levulinic-3,3,5,5,5-d5 Acid, gamma-Ketovaleric-d5 Acid
InChI formel InChI=1S/C5H8O3/c1-4(6)2-3-5(7)8/h2-3H2,1H3,(H,7,8)/i1D3,2D2
SMIL [2H]C([2H])([2H])C(=O)C([2H])([2H])CC(=O)O
IUPAC navn 3,3,5,5,5-pentadeuterio-4-oxopentanoic acid
Molekylvægt (g/mol) 121.15
Formel vægt 121.0787