Få mere at vide
Levosimendan, TRC
Artikelnummer. 30483589
Skift visning
Mængde:
10 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30483589 250 mg
30479711 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30483589 Leverandør Toronto Research Chemicals Leverandørnr. TRCL378000250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Levosimendan
CAS 141505-33-1
Renhedsgrad noter HPLC
Molekylær formel C14 H12 N6 O
Mængde 250 mg
Synonym 2-[2-[4-[(4R)-1,4,5,6-Tetrahydro-4-methyl-6-oxo-3-pyridazinyl]phenyl]hydrazinylidene]propanedinitrile, Propanedinitrile, [[4-(1,4,5,6-tetrahydro-4-methyl-6-oxo-3-pyridazinyl)phenyl]hydrazono]-, (R)-, Propanedinitrile, [[4-[(4R)-1,4,5,6-tetrahydro-4-methyl-6-oxo-3-pyridazinyl]phenyl]hydrazono]- (9CI), 2-[2-[4-[(4R)-1,4,5,6-Tetrahydro-4-methyl-6-oxo-3-pyridazinyl]phenyl]hydrazinylidene]propanedinitrile, (-)-OR 1259, (R)-Simendan, 2-[[4-[(4R)-4-Methyl-6-oxo-4,5-dihydro-1H-pyridazin-3-yl]phenyl]hydrazinylidene]propanedinitrile, Levosimendan, OR 1259, Simdax
InChI formel InChI=1S/C14H12N6O/c1-9-6-13(21)19-20-14(9)10-2-4-11(5-3-10)17-18-12(7-15)8-16/h2-5,9,17H,6H2,1H3,(H,19,21)/t9-/m1/s1
SMIL C[C@@H]1CC(=O)NN=C1c2ccc(NN=C(C#N)C#N)cc2
IUPAC navn 2-[[4-[(4R)-4-methyl-6-oxo-4,5-dihydro-1H-pyridazin-3-yl]phenyl]hydrazinylidene]propanedinitrile
Molekylvægt (g/mol) 280.28
Formel vægt 280.1073
Procent renhed >95