Få mere at vide
Levomepromazine Maleate, LoGiCal
Artikelnummer. 30463426
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30463426 10 mg
1 options
Artikelnummer. 30463426 Leverandør LoGiCal Leverandørnr. LGCFOR0019.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Levomepromazine Maleate
CAS 7104-38-3
Analyt- eller komponentnavne Levomepromazine Maleate
Forsendelsestilstand Room Temperature
Molekylær formel C19 H24 N2 O S . C4 H4 O4
Certificeringer/compliance ISO 9001
Synonym Levomepromazine Maleate
InChI formel InChI=1S/C19H24N2OS.C4H4O4/c1-14(12-20(2)3)13-21-16-7-5-6-8-18(16)23-19-10-9-15(22-4)11-17(19)21;5-3(6)1-2-4(7)8/h5-11,14H,12-13H2,1-4H3;1-2H,(H,5,6)(H,7,8)/b;2-1-/t14-;/m1./s1
Løsningstype Neat
SMIL COc1ccc2Sc3ccccc3N(C[C@H](C)CN(C)C)c2c1.OC(=O)\C=C/C(=O)O
IUPAC navn (Z)-but-2-enedioic acid;(2R)-3-(2-methoxyphenothiazin-10-yl)-N,N,2-trimethylpropan-1-amine
Industry Type Clinical Testing
Molekylvægt (g/mol) 444.54
Mængde 10 mg
Formel vægt 444.1719 g/mol