Learn More
Levofloxacin Methyl Ester, Mikromol™
Product Code. 30459770
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30459770 4 x 25 mg 100mg
30051433 25 mg 25mg
17914426 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30459770 Supplier Mikromol Supplier No. MM0846.0600254AMP
 Restricted Item
Forventet forsendelse: 30-11-2026
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Levofloxacin Methyl Ester, Mikromol
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Levofloxacin Methyl Ester
CAS 862690-19-5
Analyte or Component Names Levofloxacin Methyl Ester
Shipping Condition Room Temperature
Molecular Formula C19 H22 F N3 O4
Certifications/Compliance ISO 17025
Synonym (3S)-9-Fluoro-2,3-dihydro-3-methyl-10-(4-methyl-1-piperazinyl)-7-oxo-7H-pyrido[1,2,3-de]-1,4-benzoxazine-6-carboxylic acid methyl ester
InChI Formula InChI=1S/C19H22FN3O4/c1-11-10-27-18-15-12(17(24)13(9-23(11)15)19(25)26-3)8-14(20)16(18)22-6-4-21(2)5-7-22/h8-9,11H,4-7,10H2,1-3H3/t11-/m0/s1
Solution Type Neat
SMILES COC(=O)C1=CN2[C@@H](C)COc3c(N4CCN(C)CC4)c(F)cc(C1=O)c23
Molecular Weight (g/mol) 375.39
Quantity 4 x 25 mg
Formula Weight 375.1594 g/mol