Få mere at vide
Levofloxacin-d8, TRC
Artikelnummer. 30479709
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30479709 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30479709 Leverandør Toronto Research Chemicals Leverandørnr. TRCL3600021MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Levofloxacin-d8
CAS 1217716-71-6
Renhedsgrad noter HPLC
Molekylær formel C18H12D8FN3O4
Mængde 1 mg
Synonym (3S)-9-Fluoro-2,3-dihydro-3-methyl-10-(4-methyl-1-piperazinyl-2,2,3,3,5,5,6,6-d8)-7-oxo-7H-pyrido[1,2,3-de]-1,4-benzoxazine-6-carboxylic acid, Levofloxacin-D8 (piperazinyl-2,2,3,3,5,5,6,6-D8), Levofloxacin-d8
InChI formel InChI=1S/C18H20FN3O4/c1-10-9-26-17-14-11(16(23)12(18(24)25)8-22(10)14)7-13(19)15(17)21-5-3-20(2)4-6-21/h7-8,10H,3-6,9H2,1-2H3,(H,24,25)/t10-/m0/s1/i3D2,4D2,5D2,6D2
SMIL [2H]C1([2H])N(C)C([2H])([2H])C([2H])([2H])N(c2c(F)cc3C(=O)C(=CN4[C@@H](C)COc2c34)C(=O)O)C1([2H])[2H]
Molekylvægt (g/mol) 369.42
Procent renhed >95