Få mere at vide
Levodropropizine N-Oxide, Mikromol™
Artikelnummer. 17974346
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17974346 100 mg
17961016 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 17974346 Leverandør Mikromol Leverandørnr. MM1246.05
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Levodropropizine N-Oxide
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Levodropropizine N-Oxide
CAS 152237-40-6
Analyt- eller komponentnavne Levodropropizine N-Oxide
Forsendelsestilstand Room Temperature
Molekylær formel C13 H20 N2 O3
Certificeringer/compliance ISO 17025
Synonym 1,2-Propanediol, 3-(1-oxido-4-phenyl-1-piperazinyl)-, (S)- (9CI), 1,2-Propanediol, 3-(4-phenyl-1-piperazinyl)-, N-oxide, (S)-, Levodropropizine N-Oxide
InChI formel InChI=1S/C13H20N2O3/c16-11-13(17)10-15(18)8-6-14(7-9-15)12-4-2-1-3-5-12/h1-5,13,16-17H,6-11H2/t13-/m0/s1
SMIL OC[C@@H](O)C[N+]1([O-])CCN(CC1)c2ccccc2
IUPAC navn (2S)-3-(1-oxido-4-phenylpiperazin-1-ium-1-yl)propane-1,2-diol
Molekylvægt (g/mol) 252.31
Mængde 100 mg
Formel vægt 252.1474 g/mol