Få mere at vide
Levetiracetam-d6, TRC
Artikelnummer. 30477174
Skift visning
Mængde:
1 mg
2.5 mg
Pakningsstørrelse:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30477174 2.5 mg 2.5mg
30468698 1 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30477174 Leverandør Toronto Research Chemicals Leverandørnr. TRCL3315012.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1133229-29-4
Molekylær formel C8 2H6 H8 N2 O2
Molekylvægt (g/mol) 176.25
Synonym 1-Pyrrolidineacetamide-α-d, α-(ethyl-1,1,2,2,2-d5)-2-oxo-, (αS)-, Levetiracetam-D6
IUPAC navn (2S)-2,3,3,4,4,4-hexadeuterio-2-(2-oxopyrrolidin-1-yl)butanamide
SMIL [2H]C([2H])([2H])C([2H])([2H])[C@]([2H])(N1CCCC1=O)C(=O)N
Tekniske data
Kemisk navn eller materiale Levetiracetam butyl-D6
Mængde 2.5 mg
InChI formel InChI=1S/C8H14N2O2/c1-2-6(8(9)12)10-5-3-4-7(10)11/h6H,2-5H2,1H3,(H2,9,12)/t6-/m0/s1/i1D3,2D2,6D
Formel vægt 176.1432