Få mere at vide
Levetiracetam-d6, TRC
Artikelnummer. 30466353
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30466353 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30466353 Leverandør Toronto Research Chemicals Leverandørnr. TRCL33150310MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Levetiracetam pyrrolidin-d6
CAS 1133229-30-7
Renhedsgrad noter HPLC
Molekylær formel C8 D6 H8 N2 O2
Mængde 10 mg
Synonym (αS)-α-Ethyl-2-oxo-1-pyrrolidineacetamide-d6, (-)-Levetiracetam-d6, Keppra-d6, Levesam 500-d6, UCB-L 059-d6
InChI formel InChI=1S/C8H14N2O2/c1-2-6(8(9)12)10-5-3-4-7(10)11/h6H,2-5H2,1H3,(H2,9,12)/t6-/m0/s1/i3D2,4D2,5D2
SMIL [2H]C1([2H])N([C@@H](CC)C(=O)N)C(=O)C([2H])([2H])C1([2H])[2H]
IUPAC navn (2S)-2-(2,2,3,3,4,4-hexadeuterio-5-oxo-pyrrolidin-1-yl)butanamide
Molekylvægt (g/mol) 176.25
Formel vægt 176.1432
Procent renhed >95