Få mere at vide
Levetiracetam-d6 (2,3,3,4,4,4-butyramide-d6), CDN
Artikelnummer. 30326133
Skift visning
Mængde:
0.005 g
0.01 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30326133 0.005 g
30318014 0.01 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30326133 Leverandør CDN Leverandørnr. CDND75710.005G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Levetiracetam butyl-D6
CAS 1133229-29-4
Molekylær formel C8 2H6 H8 N2 O2
Mængde 0.005 g
Synonym 1-Pyrrolidineacetamide-α-d, α-(ethyl-1,1,2,2,2-d5)-2-oxo-, (αS)-, Levetiracetam-D6
InChI formel InChI=1 S/C8H14N2O2/c1-2-6(8(9)12)10-5-3-4-7(10)11/h6H,2-5H2,1H3,(H2,9,12)/t6-/m0/s1/i1D3,2D2,6 D
SMIL [2 H]C([2 H])([2 H])C([2 H])([2 H])[C@]([2 H])(N1CCCC1=O)C(=O)N
IUPAC navn (2 S)-2,3,3,4,4,4-hexadeuterio-2-(2-oxopyrrolidin-1-yl)butanamide
Molekylvægt (g/mol) 176.25
Formel vægt 176.1432 g/mol
Procent renhed 97 atom % D, min 98% Chemical Purity