Få mere at vide
Levetiracetam-d3 (contains d0), TRC
Artikelnummer. 30482518
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30482518 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30482518 Leverandør Toronto Research Chemicals Leverandørnr. TRCL33150450MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Levetiracetam-d3
CAS 1217851-16-5
Renhedsgrad noter HPLC
Molekylær formel C8H11D3N2O2
Mængde 50 mg
Synonym (S)-2-(2-oxopyrrolidin-1-yl)butanamide, (alpha S)-alpha -(Ethyl-D3)-2-oxo-1-pyrrolidineacetamide (-)-Levetiracetam-D3 Keppra-D3
InChI formel InChI=1S/C8H14N2O2/c1-2-6(8(9)12)10-5-3-4-7(10)11/h6H,2-5H2,1H3,(H2,9,12)/t6-/m0/s1/i1D3
SMIL O=C(N)C(N1C(=O)CCC1)CC
IUPAC navn 2S)-4,4,4-Trideuterio-2-(2-oxopyrrolidin-1-yl)butanamide
Molekylvægt (g/mol) 173.23
Formel vægt 173.12
Procent renhed >95