Få mere at vide
Levamisole EP Impurity D, Mikromol™
Artikelnummer. 30051462
Skift visning
Mængde:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30051462 25 mg
30051989 100 mg
30460894 4 x 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30051462 Leverandør Mikromol Leverandørnr. MM0706.040025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Levamisole EP Impurity D
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 6-Phenyl-2,3-dihydroimidazo[2,1-b]thiazole
CAS 4335-28-8
Analyt- eller komponentnavne 6-Phenyl-2,3-dihydroimidazo[2,1-b]thiazole
Forsendelsestilstand Room Temperature
Molekylær formel C11 H10 N2 S
Certificeringer/compliance ISO 17025
Synonym Levamisole Hydrochloride Imp. D (EP), Levamisole Imp. D (EP), 6-Phenyl-2,3-dihydroimidazo[2,1-b]thiazole, Imidazo[2,1-b]thiazole, 2,3-dihydro-6-phenyl-, 2,3-Dihydro-6-phenylimidazo[2,1-b]thiazole, Levamisole Hydrochloride Impurity D, Levamisole Impurity D
InChI formel InChI=1S/C11H10N2S/c1-2-4-9(5-3-1)10-8-13-6-7-14-11(13)12-10/h1-5,8H,6-7H2
SMIL C1Cn2cc(nc2S1)c3ccccc3
IUPAC navn 6-phenyl-2,3-dihydroimidazo[2,1-b][1,3]thiazole
Molekylvægt (g/mol) 202.28
Mængde 25 mg
Formel vægt 202.0565 g/mol