Få mere at vide
Leu-Enkephalin Amide H-Tyr-Gly-Gly-Phe-Leu-NH2 Hydrochloride Salt, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Leu-Enkephalin Amide H-Tyr-Gly-Gly-Phe-Leu-NH2 Hydrochloride Salt |
| CAS | 99236-50-7 |
| Molekylær formel | C28H38N6O6 . HCl |
| Mængde | 25 mg |
| Synonym | L-Leucinamide, L-tyrosylglycylglycyl-L-phenylalanyl-, monohydrochloride (9CI), 5-L-Leucinamide-1-5-β-neoendorphin (human) hydrochloride, L-Tyrosylglycylglycyl-L-phenylalanyl-L-leucinamide monohydrochloride, Leucine enkephalin amide hydrochloride, Leu-enkephalin amide hydrochloride |
| InChI formel | InChI=1S/C28H38N6O6.ClH/c1-17(2)12-22(26(30)38)34-28(40)23(14-18-6-4-3-5-7-18)33-25(37)16-31-24(36)15-32-27(39)21(29)13-19-8-10-20(35)11-9-19;/h3-11,17,21-23,35H,12-16,29H2,1-2H3,(H2,30,38)(H,31,36)(H,32,39)(H,33,37)(H,34,40);1H/t21-,22-,23-;/m0./s1 |
| SMIL | Cl.CC(C)C[C@H](NC(=O)[C@H](Cc1ccccc1)NC(=O)CNC(=O)CNC(=O)[C@@H](N)Cc2ccc(O)cc2)C(=O)N |
| IUPAC navn | (2S)-2-[[(2S)-2-[[2-[[2-[[(2S)-2-amino-3-(4-hydroxyphenyl)propanoyl]amino]acetyl]amino]acetyl]amino]-3-phenylpropanoyl]amino]-4-methylpentanamide;hydrochloride |
| Molekylvægt (g/mol) | 591.1 |
| Formel vægt | 590.262 |