missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Letrozole, MedChemExpress
Beskrivelse
Letrozole (CGS 20267) is a potent, selective, reversible and orally active non-steroidal inhibitor of aromatase, with an IC50 of 11.5 nM. Letrozole selective inhibits estrogen biosynthesis, and can be used for the research of breast cancer.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Letrozole |
| Fysisk form | Powder |
| CAS | 112809-51-5 |
| Farve | Off-White |
| Mængde | 500 mg |
| Renhedsgrad noter | Research |
| Molekylær formel | C17H11N5 |
| Til brug med (applikation) | Cancer-programmed cell death |
| Synonym | CGS 20267 |
| Opløselighedsinformation | DMSO : 50 mg/mL (175.25 mM; Need ultrasonic) |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?