Få mere at vide
Letrozole Diacid Impurity, TRC
Artikelnummer. 30483453
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30483453 100 mg
30477304 10 mg
30468523 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30483453 Leverandør Toronto Research Chemicals Leverandørnr. TRCL330220100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1644566-39-1
Molekylær formel C17H13N3O4
Molekylvægt (g/mol) 323.3
Synonym Benzoic acid, 4,4'-(1H-1,2,4-triazol-1-ylmethylene)bis-, 4,4'-(1H-1,2,4-Triazol-1-ylmethylene)bis[benzoic acid], 4,4'-(1H-1,2,4-Triazol-1-ylmethylene)dibenzoic Acid, 4,4'-(1H-1,2,4-triazol-1-ylmethylene)bis-benzoic Acid
IUPAC navn 4-[(4-carboxyphenyl)-(1,2,4-triazol-1-yl)methyl]benzoic acid
SMIL OC(=O)c1ccc(cc1)C(c2ccc(cc2)C(=O)O)n3cncn3
Tekniske data
Kemisk navn eller materiale Letrozole Diacid Impurity
Mængde 100 mg
InChI formel InChI=1S/C17H13N3O4/c21-16(22)13-5-1-11(2-6-13)15(20-10-18-9-19-20)12-3-7-14(8-4-12)17(23)24/h1-10,15H,(H,21,22)(H,23,24)
Formel vægt 323.0906