Få mere at vide
Letrozole-d4 (major), TRC
Artikelnummer. 30472124
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30472124 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30472124 Leverandør Toronto Research Chemicals Leverandørnr. TRCL3301021MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Letrozole-d4 (major)
CAS 1133712-96-5
Renhedsgrad noter HPLC
Molekylær formel C17D4H7N5
Mængde 1 mg
Synonym 4,4’-(1H-1,2,4-Triazol-1-ylmethylene)bisbenzonitrile-d4, 4-[1-(4-Cyanophenyl)-1-(1,2,4-triazol-1-yl)methyl]benzonitrile-d4, CGS 20267-d4, Femara-d4, Lerozole-d4, Letrazole-d4
InChI formel InChI=1S/C17H11N5/c18-9-13-1-5-15(6-2-13)17(22-12-20-11-21-22)16-7-3-14(10-19)4-8-16/h1-8,11-12,17H/i1D,2D,5D,6D
SMIL [2H]c1c([2H])c(C(c2ccc(cc2)C#N)n3cncn3)c([2H])c([2H])c1C#N
IUPAC navn 4-[(4-cyanophenyl)-(1,2,4-triazol-1-yl)methyl]-2,3,5,6-tetradeuterio-benzonitrile
Molekylvægt (g/mol) 289.33
Formel vægt 289.1266
Procent renhed >95