Få mere at vide
Lercanidipine-d3 Hydrochloride, TRC
Artikelnummer. 30467840
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30467840 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30467840 Leverandør Toronto Research Chemicals Leverandørnr. TRCL17900210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Lercanidipine-d3 Hydrochloride
CAS 1189954-18-4
Molekylær formel C36D3H38N3O6.HCl
Mængde 10 mg
Synonym 1,4-Dihydro-2,6-dimethyl-4-(3-nitrophenyl)-3,5-pyridinedicarboxylic Acid 2-[(3,3-Diphenylpropyl)(methyl-d3)amino]-1,1-dimethylethyl Methyl Ester Hydrochloride
InChI formel InChI=1S/C36H41N3O6.ClH/c1-24-31(34(40)44-6)33(28-18-13-19-29(22-28)39(42)43)32(25(2)37-24)35(41)45-36(3,4)23-38(5)21-20-30(26-14-9-7-10-15-26)27-16-11-8-12-17-27;/h7-19,22,30,33,37H,20-21,23H2,1-6H3;1H/i5D3;
SMIL Cl.[2H]C([2H])([2H])N(CCC(c1ccccc1)c2ccccc2)CC(C)(C)OC(=O)C3=C(C)NC(=C(C3c4cccc(c4)[N+](=O)[O-])C(=O)OC)C
IUPAC navn O5-[2-[3,3-diphenylpropyl(trideuteriomethyl)amino]-1,1-dimethyl-ethyl] O3-methyl 2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate;hydrochloride
Molekylvægt (g/mol) 651.21
Formel vægt 650.295