missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Leptin R Antibody (mibavademab), HRP, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals NBP3-28039H
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Leptin R Monoclonal antibody specifically detects Leptin R in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Tekniske data
| Leptin R | |
| Monoclonal | |
| HRP | |
| CD295 antigen, DB, DKFZp686B1731, huB219, LEP-R, leptin receptor, OB receptor, OB-R, OBRCD295 | |
| LEPR/ CD295 | |
| 0.1 mL | |
| Signal Transduction | |
| 3953 | |
| Store at 4°C in the dark. | |
| IgG4 |
| Flow Cytometry, ELISA, Functional Assay | |
| mibavademab | |
| PBS | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion