missing translation for 'onlineSavingsMsg'
Få mere at vide

Leonurine, MedChemExpress

Artikelnummer. 16489078
Encompass_Preferred
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
5 mg
10 mg
25 mg
50 mg
100 mg
Pakningsstørrelse:
100mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
16489078 10 mg -
16474877 50 mg -
16433896 25 mg -
16429136 100 mg 100mg
16411498 5 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16489078 Supplier Medchem Express Supplier No. HYN0741

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

This item is not returnable. View return policy

MedChemExpress Leonurine is an alkaloid isolated from Herba leonuri, with anti-oxidative and anti-inflammatory.

Leonurine is an alkaloid isolated from Herba leonuri, with anti-oxidative and anti-inflammatory.

Specifications

Kemisk navn eller materiale Leonurine
CAS 24697-74-3
Farve White
Renhedsgrad noter Research
Molekylær formel C14H21N3O5
Mængde 10 mg
Synonym SCM-198
Opløselighedsinformation DMSO : 33.33 mg/mL (107.06 mM; ultrasonic and adjust pH to 2 with HCl) ∣H2O : < 0.1 mg/mL (insoluble)
SMIL O=C(OCCCCNC(N)=N)C1=CC(OC)=C(O)C(OC)=C1
Molekylvægt (g/mol) 311.33
Formel vægt 311.33
Procent renhed 99.61%
Grad Research
Fysisk form Solid
Til brug med (applikation) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.