Få mere at vide
Tocris Bioscience™ Lenalidomide 4'-alkylC5-azide
Beskrivelse
Lenalidomide 4'-alkylC5-azide is a functionalized cereblon ligand for PROTAC® research and development; incorporates an E3 ligase ligand plus an alkylC5 linker with terminal azide ready for conjugation to a target protein ligand. Part of a range of functionalized tool molecules for PROTAC R&D.
PROTAC® is a registered trademark of Arvinas Operations, Inc., and is used under license.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 3-[4-[(5-Azidopentyl)amino]-1,3-dihydro-1-oxo-2 H-isoindol-2-yl]-2,6-piperidinedione |
| CAS | 2399455-78-6 |
| Mængde | 25 mg |
| Mål | Degrader Building Blocks |
| Molekylær formel | C18H22N6O3 |
| Renhed | 0.95 |