Få mere at vide
Tocris Bioscience™ LEI 401
Beskrivelse
LEI 401 is a high affinity N-acylphosphatidylethanolamine phospholipase D (NAPE-PLD) inhibitor (Ki = 0.027 and 0.18 μM for human and mouse NAPE-PLD, respectively; IC50 for human enzyme = 0.86 μM). LEI 401 exhibits no significant activity at cannabinoid receptors or other metabolic enzymes of the endocannabinoid system. The product reduces levels of N-acylethanolamines (NAE) in a neuroblastoma cell line in vitro. It also reduces anandamide levels in mouse brain in vivo and impairs fear extinction. LEI 701 is CNS penetrant and cell-permeable.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | N-(Cyclopropylmethyl)-6-[(3 S)-3-hydroxy-1-pyrrolidinyl]-2-[(3 S)-3-phenyl-1-piperidinyl]-4-pyrimidinecarboxamide |
| CAS | 2393840-15-6 |
| Mængde | 10 mg |
| Mål | Phospholipase Inhibitors |
| Molekylær formel | C24H31N5O2 |
| Renhed | 0.98 |