Learn More
BINDER™ Left Access Port (50 to 125 mm)
For Use With: BINDER units series BD
Brand: BINDER™ 8012-0051
This item is not returnable.
View return policy
Description
- For use on left
- Includes silicone plug
- For use with a wide range of BINDER™ incubators and chambers
Specifications
Access port with silicone plug - 50 mm, left | |
Steel | |
Left | |
50 mm |
BINDER Chambers Series BD | |
Access Port | |
Silicone Plug |
Product Content Correction
Product Title
BINDER™ Left Access Port (50 to 125 mm) > Diameter: 50 mm, For Use With: Series BD
Spot an opportunity for improvement?Share a Content Correction