Learn More
Syngene™ LED Module for Syngene Gbox Chemi System
Syngene™ LED Module for Syngene Gbox Chemi System for western fluorescence
Brand: Syngene™ LED-BLUE x 2
Additional Details : Weight : 0.02000kg
Specifications
Blue | |
Gbox Chemi Systems | |
LED Module |
2 | |
465nm |