Learn More
Lead solution for ICP/MS 10mg/L HNO3 matrix, Fisher chemical™
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
430.00 DKK - 785.00 DKK
Chemical Identifiers
CAS | 7697-37-2 |
---|---|
Molecular Formula | HNO3 |
Molecular Weight (g/mol) | 63.01 |
MDL Number | MFCD00011349 |
InChI Key | GRYLNZFGIOXLOG-UHFFFAOYSA-N |
PubChem CID | 944 |
ChEBI | CHEBI:48107 |
IUPAC Name | nitric acid |
SMILES | O[N+]([O-])=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
12967393
|
Fisher Chemical
S 641 050 |
50 mL | Bottle |
785.00 DKK
50mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12977393
![]() |
Fisher Chemical
S 641 100 |
100 mL | Bottle |
430.00 DKK
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Chemical Identifiers
7697-37-2 | |
63.01 | |
GRYLNZFGIOXLOG-UHFFFAOYSA-N | |
CHEBI:48107 | |
O[N+]([O-])=O |
HNO3 | |
MFCD00011349 | |
944 | |
nitric acid |
Specifications
7697-37-2 | |
10 mg/L | |
GRYLNZFGIOXLOG-UHFFFAOYSA-N | |
nitric acid | |
63.01 | |
CHEBI:48107 |
HNO3 | |
MFCD00011349 | |
O[N+]([O-])=O | |
HNO3 | |
944 | |
Lead |
Safety and Handling
missing translation for 'shelfLife' : 12 months
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Lead solution for ICP/MS 10mg/L HNO3 matrix, Fisher chemical™