Learn More
Lead Solution for ICP, 1g/L, HNO3 Matrix, Fisher Chemical™
Catalog Number 12903474
Click to view available options
Quantity:
100 mL
250 mL
500 mL
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
Concentration 1g/L
Quantity 100 mL
Solution Contains HNO3