Få mere at vide
STERIS™ Lead-free Autoclave Indicator Tape
Artikelnummer. p-8014775
Dimensioner (LxB):
50 m x 19 mm
50 m x 25 mm
Pakningsstørrelse:
Hver
Artikelnummer. 12370489
Brand: STERIS™ 0162AB
Seal products and packages for sterilization with this self-adhering tape. STERIS™ Lead-free Autoclave Indicator Tape features green lead-free stripes that change color to brown/black providing an immediate indication that the sterilization process is complete.
  • Self-adhering tape with a chemical indicator printed on one side
  • The easily recognizable stripes change color when exposed to the specific sterilization, verifying exposure to the process without the need to open packs or check records
  • Color change brown/black
TRUSTED_SUSTAINABILITY
Tekniske data
Farve Brown, Black
Dimensioner (LxB) 50 m x 19 mm