Få mere at vide
Brand™ LDPE Wash Bottles without Valve
Shop alle Brand produkter
Kapacitet (metrisk):
250 mL
500 mL
Kemisk etiket:
Acetone
Distilled Water
Ethanol
Isopropanol
Methanol
Neutral
Lukningsfarve:
Blue
Green
Orange
Red
White
Yellow
Pakningsstørrelse:
Pakke med 5
Beskrivelse
- Manufactured from flexible LDPE
- Wide-mouth.
- High-quality wash bottles with integrated wash stream and sealing cone closure.
- Large opening for convenient filling.
- Label with designation of contents and general safety statements in English (with CAS numbers and NFPA codes) and German (with CAS numbers).
Tekniske data
Tekniske data
| Kapacitet (metrisk) | 500 mL |
| Materiale | LDPE |
| Lukningsfarve | Yellow |
| Mund | Wide |
| Produkttype | Wash Bottle |
| Kemisk etiket | Neutral |
| Lukketype | Sealing Cone |