Learn More
Karl Hecht™ LDPE Dropping Bottles
Product Code. 10084111
Change view
Click to view available options
Capacity (Metric):
250 mL
Closure Size:
GL 25
Unit Size:
Each
Product Code. Capacity (Metric) Closure Size Unit Size
10084111 250 mL GL 25 Each
1 options
Product Code. 10084111 Supplier Karl Hecht™ Supplier No. 49103024
  • ONLINE EXCLUSIVE PRICE
    20.77 DKK
    12.01 DKK / Pakke med 1
    Spare 8.76 DKK
    42% Slukket
View all promotions
 Restricted Item
Forventet forsendelse: 18-12-2026
Provided with a dropping tip for precise and repeatable dispensing of liquids.
  • Made of polyethylene
  • With attached cap
  • Available in a range of capacities and thread sizes
Specifications
Capacity (Metric) 250 mL
Material LDPE
Closure Color Red
Closure Included Yes
Closure Size GL 25
Closure Type Attached Cap
Product Type Dropping Bottle