missing translation for 'onlineSavingsMsg'
Få mere at vide

STEMCELL Technologies LDN193189 (Dihydrochloride)

Artikelnummer. 17130032
Encompass_Preferred
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17130032 50 mg
17120032 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 17130032 Leverandør STEMCELL Technologies Leverandørnr. 72149

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

BMP pathway inhibitor; Inhibits ALK1, ALK2, ALK3, and ALK6

  • LDN193189 is a potent inhibitor of the bone morphogenetic (BMP) pathway, inhibiting ALK1, ALK2, ALK3, and ALK6 (IC50 = 0.8, 0.8, 5.3, and 16.7 nM respectively; Sanvitale et al.) It is a derivative of Dorsomorphin (Catalog #72102) that is typically used at approximately 100-fold lower concentrations (Sanvitale et al.; Vogt et al.) This product is supplied as the dihydrochloride salt of the molecule and it is the most soluble version of LDN193189
  • Promotes differentiation of neural progenitor cells from human pluripotent stem cells (Chambers et al.; Kriks et al.)
  • Promotes differentiation of neural crest cells from human pluripotent stem cells (Kreitzer et al.)
  • Promotes differentiation of anterior foregut endoderm from human and mouse pluripotent stem cell-derived definitive endoderm (Kearns et al.)
  • Promotes differentiation of sensory epithelial cells of the inner ear from mouse embryonic stem cells (Koehler et al.)
  • Inhibits prostate and breast cancer tumor growth (Balboni et al.; Lee et al.)
  • Prevents ovarian cancer cell proliferation (Tsai et al.)

Tekniske data

CAS 1435934-00-1
Synonym DM-3189
Mængde 50 mg
Molekylær formel C25H22N6 · 2HCl
Renhed >98%
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.