Beskrivelse
LC3 Antibody Packantibody specifically detects LC3 in Human, Mouse, Rat, Porcine, Alligator, Amphibian, Avian, Bacteria, Bovine, Canine, Chicken, Chinese Hamster, Fish, Guinea Pig, Hamster, Invertebrate, Plant, Primate, Monkey, Rabbit, Golden Syrian Hamster, Zebrafish samples. It is validated for Western Blot, Simple Western, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoblot, Proximity Ligation Assay, SDS-Page, Southern Blot, Immunohistochemistry (Whole-Mount), ChIP assay (ChIP), Gene Knock-Out, KnockDown
Tekniske data
Tekniske data
| Antigen | LC3 |
| Applikationer | Western Blot, Simple Western, Flow Cytometry, ELISA, Immunohistochemistry |
| Konjugeret | Unconjugated |
| Gene Alias | ATG8F, LC3B, MAP1A/1BLC3, map1lc3b, MAP1LC3B-a, microtubule associated protein 1 light chain 3 beta |
| Immunogen | For NB100-2331: A synthetic peptide made to an internal portion of the human LC3 protein sequence (between residues 25-121) [Uniprot: Q9H492]., For NB100-2220: Polyclonal LC3B Antibody was made to a synthetic peptide made to an N-terminal portion of the human LC3B protein sequence (between residues 1-100) [UniProt# Q9GZQ8]. |
| Mængde | 1 Pack |
| Regulatorisk status | RUO |
| Forskningsdisciplin | Autophagy, Cancer, Cardiovascular Biology, mTOR Pathway |
| Gen-id (Entrez) | 81631 |
| Målarter | Human, Mouse, Rat, Pig, Alligator |
| Vis mere |