missing translation for 'onlineSavingsMsg'
Få mere at vide

Lauryl Betaine, TRC

Artikelnummer. 30180352
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30180352 1 g
30180237 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.
Artikelnummer. 30180352 Leverandør Toronto Research Chemicals Leverandørnr. TRCL1787501G

Please to purchase this item. Need a web account? Register with us today!

Denne vare kræver en prisopdatering, inden den kan bestilles.

Anmod om en pris på denne vare her.

Denne vare kan ikke returneres. Se returpolicy
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 683-10-3
Molekylær formel C16 H33 N O2
Molekylvægt (g/mol) 271.44
Synonym 1-Dodecanaminium, N-(carboxymethyl)-N,N-dimethyl-, inner salt, 1-Dodecanaminium, N-(carboxymethyl)-N,N-dimethyl-, hydroxide, inner salt, Ammonium, (carboxymethyl)dodecyldimethyl-, hydroxide, inner salt (7CI,8CI), Glycine, dodecyldimethylbetaine (6CI), (Carboxymethyl)dodecyldimethylammonium hydroxide inner salt, (Dodecyldimethylammonio)acetate, (Dodecyldimethylammonio)ethanoate, AM 30, AM 301, Adeka Amphote AB 35L, Adeka Amphote PB 30L, Ambiteric D 40, Amigen S, Amipol 6S, Amogen S, Amogen S-H, Amphitol 20BS, Amphitol 24B, Anfoterico LB, Anon BL, Anon BL-SF, BS 12, BS 12 (betaine surfactant), Betadet DM 20, Betaine lauryldimethylaminoacetate, Bister ML, Culveram CDG, DDN, Desimex i, Dimethylaurylbetaine, Dimethyldodecylbetaine, Dimethyllaurylaminoacetate betaine, Dimethyllaurylbetaine, Dodecylbetaine, Dodecyldimethylbetaine, Empigen BB/L, Genagen B 1566J, Genagen LAB, Lauryl-N,N-dimethylacetic acid betaine, Lauryl-N-betaine, Lauryl-N-methylsarcosine, Laurylbetain, Laurylbetaine, Lauryldimethylaminoacetate betaine, Lauryldimethylaminoacetic acid betaine, Lauryldimethylaminoacetic betaine, Lauryldimethylammonioacetate, Lauryldimethylbetaine, Lebon LD 36, Lonzaine 12S, Mackam LB 35, Makam LB 35, N,N-Dimethyl-N-dodecylglycine betaine, N,N-Dimethyl-N-laurylglycine inner salt, N,N-Dimethyldodecylbetaine, N-(Carboxymethyl)-N-lauryldimethylammonium hydroxide inner salt, N-Carboxymethyl-N,N-dimethyl-1-dodecanaminium inner salt, N-Dodecyl-N,N-dimethylbetaine, N-Lauryl-N,N-dimethyl-N-(carboxymethyl)ammonium carbobetaine, N-Lauryl-N,N-dimethyl-α-betaine, N-Lauryl-N,N-dimethylaminomethanecarboxylate inner salt, Nikkol AM 301, Nissan Anon BL, Nissan Anon BL-SF, Obazoline LB, Obazoline LB-SF, Pionin C 157K, Product DDN, Rewoteric AM-DML, Rewoteric AM-DML 35, Rikabion A 100, Swanol AM 301, Synthecol BT, TC, TC (antistatic agent), Takesurf C 157L, Varion CDG-K, α-(Dodecyldimethylammonio)-ω-acetate
IUPAC navn 2-[dodecyl(dimethyl)azaniumyl]acetate
SMIL CCCCCCCCCCCC[N+](C)(C)CC(=O)[O-]

Tekniske data

Kemisk navn eller materiale Lauryl Betaine
Mængde 1 g
InChI formel InChI=1S/C16H33NO2/c1-4-5-6-7-8-9-10-11-12-13-14-17(2,3)15-16(18)19/h4-15H2,1-3H3
Formel vægt 271.2511
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.