Få mere at vide
Lauroyl-d3-L-carnitine Hydrochloride, TRC
Artikelnummer. 30469830
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30469830 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30469830 Leverandør Toronto Research Chemicals Leverandørnr. TRCL17868325MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Lauroyl-d3-L-carnitine Hydrochloride
CAS 1297271-52-3
Molekylær formel C19H35D3NO4 . Cl
Mængde 25 mg
Synonym (2R)-3-Carboxy-N,N,N-trimethyl-2-[(1-oxododecyl-d3)oxy]-1-propanaminium Chloride
InChI formel InChI=1S/C19H37NO4.ClH/c1-5-6-7-8-9-10-11-12-13-14-19(23)24-17(15-18(21)22)16-20(2,3)4;/h17H,5-16H2,1-4H3;1H/t17-;/m1./s1/i1D3;
SMIL O=C(CCCCCCCCCCC([2H])([2H])[2H])O[C@H](CC(O)=O)C[N+](C)(C)C.[Cl-]
IUPAC navn (R)-3-carboxy-2-((dodecanoyl-12,12,12-d3)oxy)-N,N,N-trimethylpropan-1-aminium chloride
Molekylvægt (g/mol) 382.98
Formel vægt 382.27