Få mere at vide
Lauric Acid-d3, TRC
Artikelnummer. 30483839
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30483839 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30483839 Leverandør Toronto Research Chemicals Leverandørnr. TRCL17855410MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Lauric Acid-d3
CAS 79050-22-9
Molekylær formel C12 D3 H21 O2
Mængde 10 mg
Synonym Dodecanoic-12,12,12-d3 Acid, Lauric-12,12,12-d3 Acid, Dodecanoic-12,12,12-d3 acid (9CI), Lauric-12,12,12-d3 acid (6CI), [12,12,12-2H3]Dodecanoic acid, Dodecanoic acid-12,12,12-d3, Lauric acid-12,12,12-d3
InChI formel InChI=1S/C12H24O2/c1-2-3-4-5-6-7-8-9-10-11-12(13)14/h2-11H2,1H3,(H,13,14)/i1D3
SMIL [2H]C([2H])([2H])CCCCCCCCCCC(=O)O
IUPAC navn 12,12,12-trideuteriododecanoic acid
Molekylvægt (g/mol) 203.34
Formel vægt 203.1965