Learn More
Lauraldehyde, Sigma-Aldrich™
Specifications
Specifications
| CAS | 112-54-9 |
| Melting Point | 11°C to 13°C |
| Density | 0.83 g/cm3 at 20°C |
| Boiling Point | 237°C to 239°C/1013 hPa |
| Molecular Formula | CH3(CH2)10CHO |
| MDL Number | MFCD00007017 |
| Quantity | 100 mL |
| Synonym | Lauraldehyde, Dodecanal, Dodecylaldehyde, Lauraldehyde |
| Molecular Weight (g/mol) | 184.32 |
| Grade | Research |