Få mere at vide
Lathosterol, TRC
Artikelnummer. 30010990
Skift visning
Mængde:
10 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30010990 25 mg
30012878 100 mg
30011759 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30010990 Leverandør Toronto Research Chemicals Leverandørnr. TRCL17750025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 80-99-9
Molekylær formel C27 H46 O
Molekylvægt (g/mol) 386.65
Synonym Cholest-7-en-3-ol, (3β,5α)- (9CI, ACI), (3β,5α)-Cholest-7-en-3-ol (ACI), 5α-Cholest-7-en-3β-ol (8CI), 3β-Hydroxy-5α-cholest-7-ene, 7-En-3-cholestanol, Cholest-7-en-3-ol, Lathosterol, γ-Cholestenol, δ7-Cholestenol
IUPAC navn (3S,5S,9R,10S,13R,14R,17R)-10,13-dimethyl-17-[(2R)-6-methylheptan-2-yl]-2,3,4,5,6,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-3-ol
SMIL CC(C)CCC[C@@H](C)[C@H]1CC[C@H]2C3=CC[C@H]4C[C@@H](O)CC[C@]4(C)[C@H]3CC[C@]12C
Tekniske data
Kemisk navn eller materiale Lathosterol
Renhedsgrad noter HPLC
Mængde 25 mg
InChI formel InChI=1S/C27H46O/c1-18(2)7-6-8-19(3)23-11-12-24-22-10-9-20-17-21(28)13-15-26(20,4)25(22)14-16-27(23,24)5/h10,18-21,23-25,28H,6-9,11-17H2,1-5H3/t19-,20+,21+,23-,24+,25+,26+,27-/m1/s1
Formel vægt 386.3549
Procent renhed >95%